Analytical standard, Natamycin, reference material, Honeywell Fluka™

Supplier: Honeywell Chemicals

R1116-50MG
HONCR1116-50MGEA 194 SGD
HONCR1116-50MG
Analytical standard, Natamycin, reference material, Honeywell Fluka™
Standards Organic Standards

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results

Order Now

SPECIFICATIONS

Description Natamycin
Environmentally Preferable
CAS No. 7681-93-8
Formula C₃₃H₄₇NO₁₃
Melting point 280 °C
Storage temperature 2…8 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR