Pesticide reference standard, Aminocyclopyrachlor, neat, Honeywell Fluka™

R1117-25MG
HONCR1117-25MGEA 129 SGD
HONCR1117-25MG
Pesticide reference standard, Aminocyclopyrachlor, neat, Honeywell Fluka™
Standards Organic Standards

High quality inorganic and organic reference materials produced according to ISO 9001.


  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results

Order Now

SPECIFICATIONS

Description Aminocyclopyrachlor
Environmentally Preferable
CAS No. 858956-08-8
Formula C₈H₈ClN₃O₂
Storage temperature Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR