Organic analytical standards, Ethyl 4-hydroxybenzoate-ring-13C6 solution, 10 μg/ml in acetone, reference material, Honeywell Fluka™

Supplier: Honeywell Chemicals

R1190-5ML
HONCR1190-5MLEA 733 SGD
HONCR1190-5ML
Organic analytical standards, Ethyl 4-hydroxybenzoate-ring-13C6 solution, 10 μg/ml in acetone, reference material, Honeywell Fluka™
Standards Organic Standards

High quality inorganic and organic reference materials produced according to ISO 9001.


  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results

Order Now

SPECIFICATIONS

Description Ethyl 4-hydroxybenzoate-ring-¹³C₆ solution
Environmentally Preferable
Formula ¹³C₆C₃H₁₀O₃
Freezing point –95 °C
Boiling point 55...57 °C
Flash point –17 °C
Storage temperature –20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR