Analytical standard, Parbendazole, reference material, Honeywell Fluka™

Supplier: Honeywell Chemicals

R1196-25MG
HONCR1196-25MGEA 145 SGD
HONCR1196-25MG
Analytical standard, Parbendazole, reference material, Honeywell Fluka™
Standards Organic Standards

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results

Order Now

SPECIFICATIONS

Description Parbendazole
Environmentally Preferable
CAS No. 14255-87-9
Formula C₁₃H₁₇N₃O₂
Melting point 255…257 °C
Storage temperature 2…8 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR