Analytical standard, Desoxycarbadox, reference material, Honeywell Fluka™

Supplier: Honeywell Chemicals

R1207-10MG
HONCR1207-10MGEA 335 SGD
HONCR1207-10MG
Analytical standard, Desoxycarbadox, reference material, Honeywell Fluka™
Standards Organic Standards

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results

Order Now

SPECIFICATIONS

Description Desoxycarbadox
Environmentally Preferable
CAS No. 55456-55-8
Formula C₁₁H₁₀N₄O₂
Melting point 245 °C
Storage temperature Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR