Analytical standard, Zilpaterol hydrochloride, reference material, Honeywell Fluka™

Supplier: Honeywell Chemicals

R1230-10MG
HONCR1230-10MGEA 1300 SGD
HONCR1230-10MG
Analytical standard, Zilpaterol hydrochloride, reference material, Honeywell Fluka™
Standards Organic Standards

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results

Order Now

SPECIFICATIONS

Description Zilpaterol hydrochloride
Environmentally Preferable
CAS No. 119520-06-8
Formula C₁₄H₁₉N₃O₂ · HCl
Melting point 229…233 °C
Storage temperature 2…8 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR