Analytical standard, Trifloxystrobin Metabolite CGA 321113, reference material, Honeywell Fluka™

Supplier: Honeywell Chemicals

R1275-10MG
HONCR1275-10MGEA 442 SGD
HONCR1275-10MG
Analytical standard, Trifloxystrobin Metabolite CGA 321113, reference material, Honeywell Fluka™
Standards Organic Standards

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results

Order Now

SPECIFICATIONS

Description Trifloxystrobin Metabolite CGA 321113
Environmentally Preferable
CAS No. 252913-85-2
Formula C₁₉H₁₇F₃N₂O₄
Melting point 114…116 °C
Storage temperature Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR