Analytical standard, Dichlorprop-isooctyl, reference material, Honeywell Fluka™

Supplier: Honeywell Chemicals

R1300-25MG
HONCR1300-25MGEA 703 SGD
HONCR1300-25MG
Analytical standard, Dichlorprop-isooctyl, reference material, Honeywell Fluka™
Standards Organic Standards

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results

Order Now

SPECIFICATIONS

Description Dichlorprop-isooctyl
Environmentally Preferable
CAS No. 28631-35-8
Formula C₁₇H₂₄Cl₂O₃
Storage temperature Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR