Analytical standard, Phenmedipham-(methoxy-d₃), reference material, Honeywell Fluka™
Supplier: Honeywell Chemicals
R1302-10MG
HONCR1302-10MGEA 1270 SGD
HONCR1302-10MG
Analytical standard, Phenmedipham-(methoxy-d₃), reference material, Honeywell Fluka™
Standards Organic Standards
  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results