Pharmaceutical analytical standard, Terconazole, reference material, Honeywell Fluka™

Supplier: Honeywell Chemicals

R1332-100MG
HONCR1332-100MGEA 111 SGD
HONCR1332-100MG
Pharmaceutical analytical standard, Terconazole, reference material, Honeywell Fluka™
Standards Organic Standards

High quality inorganic and organic reference materials produced according to ISO 9001.


  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results

Order Now

SPECIFICATIONS

Description Terconazole
Environmentally Preferable
CAS No. 67915-31-5
Formula C₂₆H₃₁Cl₂N₅O₃
Freezing point 126...128 °C
Storage temperature Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR