Pesticide reference standard, Isofenphos solution, 100 μg/ml in acetonitrile, Honeywell Fluka™

Supplier: Honeywell Chemicals

R1361-2ML
HONCR1361-2MLEA 169 SGD
HONCR1361-2ML
Pesticide reference standard, Isofenphos solution, 100 μg/ml in acetonitrile, Honeywell Fluka™
Standards Organic Standards

High quality inorganic and organic reference materials produced according to ISO 9001.


  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results

Order Now

SPECIFICATIONS

Description Isofenphos solution
Environmentally Preferable
Formula C₁₅H₂₄NO₄PS
Boiling point 82 °C
Flash point 6 °C
Storage temperature 2...8 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR