Pesticide reference standard, Pentanochlor solution, 100 μg/ml in acetonitrile, Honeywell Fluka™
Supplier: Honeywell Chemicals
R1362-2ML
HONCR1362-2MLEA 276 SGD
HONCR1362-2ML
Pesticide reference standard, Pentanochlor solution, 100 μg/ml in acetonitrile, Honeywell Fluka™
Standards Organic Standards
High quality inorganic and organic reference materials produced according to ISO 9001.
  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results