Pesticide reference standard, Flonicamid Metabolite TFNA, neat, Honeywell Fluka™

Supplier: Honeywell Chemicals

R1373-25MG
HONCR1373-25MGEA 301 SGD
HONCR1373-25MG
Pesticide reference standard, Flonicamid Metabolite TFNA, neat, Honeywell Fluka™
Standards Organic Standards

High quality inorganic and organic reference materials produced according to ISO 9001.


  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results

Order Now

SPECIFICATIONS

Description Flonicamid Metabolite TFNA
Environmentally Preferable
CAS No. 158063-66-2
Formula C₇H₄F₃NO₂
Freezing point 133...137 °C
Storage temperature Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR