ASTM® D6584 Monoolein solution, 5000 μg/ml in pyridine, analytical standard, Honeywell Fluka™
Supplier: Honeywell Chemicals
R1381-3ML
HONCR1381-3MLEA 105 SGD
HONCR1381-3ML
ASTM® D6584 Monoolein solution, 5000 μg/ml in pyridine, analytical standard, Honeywell Fluka™
Standards Petroleum Industry Standards
High quality inorganic and organic reference materials produced according to ISO 9001.
  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results