ASTM® D6584 individual stock and internal standards kit, analytical standard, Honeywell Fluka™
Supplier: Honeywell Chemicals
R1392-1EA
HONCR1392-1EAEA 1150 SGD
HONCR1392-1EA
ASTM® D6584 individual stock and internal standards kit, analytical standard, Honeywell Fluka™
Standards Petroleum Industry Standards
High quality inorganic and organic reference materials produced according to ISO 9001.
  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
Kit contains:
R1387-1ML ASTM® D6584 Standard Solution 1, in pyridine, reference material
R1388-1ML ASTM® D6584 Standard Solution 2, in pyridine, reference material
R1389-1ML ASTM® D6584 Standard Solution 3, in pyridine, reference material
R1390-1ML ASTM® D6584 Standard Solution 4, in pyridine, reference material
R1391-1ML ASTM® D6584 Standard Solution 5, in pyridine, reference material
R1384-5ML ASTM® D6584 Butanetriol solution, 1000 μg/ml in pyridine, reference material
R1385-5ML ASTM® D6584 Tricaprin solution, 8000 μg/ml in pyridine, reference material