Pharmaceutical analytical standard, Sotalol hydrochloride, reference material, Honeywell Fluka™

Supplier: Honeywell Chemicals

R1509-10MG
HONCR1509-10MGEA 204 SGD
HONCR1509-10MG
Pharmaceutical analytical standard, Sotalol hydrochloride, reference material, Honeywell Fluka™
Standards Organic Standards

High quality inorganic and organic reference materials produced according to ISO 9001.


  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results

Order Now

SPECIFICATIONS

Description Sotalol hydrochloride
Environmentally Preferable
CAS No. 959-24-0
Formula C₁₂H₂₀N₂O₃S · HCl
Freezing point 204...207 °C
Storage temperature Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR