Pesticide reference standard, (S,S)-Metalaxyl Metabolite CGA 226048, neat, Honeywell Fluka™

R1522-10MG
HONCR1522-10MGEA 154 SGD
HONCR1522-10MG
Pesticide reference standard, (S,S)-Metalaxyl Metabolite CGA 226048, neat, Honeywell Fluka™
Standards Organic Standards

High quality inorganic and organic reference materials produced according to ISO 9001.


  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results

Order Now

SPECIFICATIONS

Description (S,S)-Metalaxyl Metabolite CGA 226048
Environmentally Preferable
Formula C₁₈H₂₅NO₆
Freezing point 92...95 °C
Storage temperature Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR