Pesticide reference standard, Fenthion-oxon, neat, Honeywell Fluka™

Supplier: Honeywell Chemicals

R1529-10MG
HONCR1529-10MGEA 595 SGD
HONCR1529-10MG
Pesticide reference standard, Fenthion-oxon, neat, Honeywell Fluka™
Standards Organic Standards

High quality inorganic and organic reference materials produced according to ISO 9001.


  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results

Order Now

SPECIFICATIONS

Description Fenthion-oxon
Environmentally Preferable
CAS No. 6552-12-1
Formula C₁₀H₁₅O₄PS
Storage temperature Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR