Organic analytical standards, Cyclohexane, reference material, Honeywell Fluka™

Supplier: Honeywell Chemicals

R1584-5ML
HONCR1584-5MLEA 59.8 SGD
HONCR1584-5ML
Organic analytical standards, Cyclohexane, reference material, Honeywell Fluka™
Standards Organic Standards

High quality organic reference materials produced according to ISO 9001.


Delivery information: Supplied with complete Certificate of Analysis (CoA) with expiry date and lot specific analytical results.

Order Now

SPECIFICATIONS

Description Cyclohexane
Environmentally Preferable
CAS No. 110-82-7
Formula C₆H₁₂
Storage temperature Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR