Organic analytical standards, n-Hexane, reference material, Honeywell Fluka™
Supplier: Honeywell Chemicals
R1585-5ML
HONCR1585-5MLEA 228 SGD
HONCR1585-5ML
Organic analytical standards, n-Hexane, reference material, Honeywell Fluka™
Standards Organic Standards
High quality organic reference materials produced according to ISO 9001.
Delivery information: Supplied with complete Certificate of Analysis (CoA) with expiry date and lot specific analytical results.