Organic analytical standards, n-Propylacetate, reference material, Honeywell Fluka™

Supplier: Honeywell Chemicals

R1594-5ML
HONCR1594-5MLEA 219 SGD
HONCR1594-5ML
Organic analytical standards, n-Propylacetate, reference material, Honeywell Fluka™
Standards Organic Standards

High quality organic reference materials produced according to ISO 9001.


Delivery information: Supplied with complete Certificate of Analysis (CoA) with expiry date and lot specific analytical results.

Order Now

SPECIFICATIONS

Description n-Propylacetate
Environmentally Preferable
CAS No. 109-60-4
Formula CH₃COOCH₂CH₂CH₃
Storage temperature Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR