Organic analytical standards, 2-Methoxyethanol, reference material, Honeywell Fluka™

Supplier: Honeywell Chemicals

R1598-5ML
HONCR1598-5MLEA 351 SGD
HONCR1598-5ML
Organic analytical standards, 2-Methoxyethanol, reference material, Honeywell Fluka™
Standards Organic Standards

High quality organic reference materials produced according to ISO 9001.


Delivery information: Supplied with complete Certificate of Analysis (CoA) with expiry date and lot specific analytical results.

Order Now

SPECIFICATIONS

Description 2-Methoxyethanol
Environmentally Preferable
CAS No. 109-86-4
Formula CH₃OCH₂CH₂OH
Storage temperature Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR