Pesticide standard, Indoxacarb, reference material, Honeywell Fluka™

Supplier: Honeywell Chemicals

R1631-25MG
HONCR1631-25MGEA 229 SGD
HONCR1631-25MG
Pesticide standard, Indoxacarb, reference material, Honeywell Fluka™
Standards Organic Standards

High quality organic reference materials produced according to ISO 9001.


Delivery information: Supplied with complete Certificate of Analysis (CoA) with expiry date and lot specific analytical results.

Order Now

SPECIFICATIONS

Description Indoxacarb
Environmentally Preferable
CAS No. 144171-61-9
Formula C₂₂H₁₇ClF₃N₃O₇
Storage temperature Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR