Pharmaceutical analytical standard, 2-NP-AHD, reference material, Honeywell Fluka™

Supplier: Honeywell Chemicals

R1657-10MG
HONCR1657-10MGEA 378 SGD
HONCR1657-10MG
Pharmaceutical analytical standard, 2-NP-AHD, reference material, Honeywell Fluka™
Standards Organic Standards

High quality organic reference materials produced according to ISO 9001.


Delivery information: Supplied with complete Certificate of Analysis (CoA) with expiry date and lot specific analytical results.

Order Now

SPECIFICATIONS

Description 2-NP-AHd
Environmentally Preferable
CAS No. 623145-57-3
Formula C₁₀H₈N₄O₄
Storage temperature Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR