Pesticide standard, Metosulam, reference material, Honeywell Fluka™

Supplier: Honeywell Chemicals

R1689-100MG
HONCR1689-100MGEA 298 SGD
HONCR1689-100MG
Pesticide standard, Metosulam, reference material, Honeywell Fluka™
Standards Organic Standards

High quality organic reference materials produced according to ISO 9001.


Delivery information: Supplied with complete Certificate of Analysis (CoA) with expiry date and lot specific analytical results.

Order Now

SPECIFICATIONS

Description Metosulam
Environmentally Preferable
CAS No. 139528-85-1
Formula C₁₄H₁₃Cl₂N₅O₄S
Storage temperature Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR