Silicon dioxide ~99% 0.5-10 μm (∼ 80% 1-5 μm), Fluka™

Supplier: Honeywell Chemicals
Danger

S5631-100G S5631-1KG S5631-500G
HONCS5631-100GEA 512 SGD
HONCS5631-100G HONCS5631-1KG HONCS5631-500G
Silicon dioxide ~99% 0.5-10 μm (∼ 80% 1-5 μm), Fluka™
Silicium dioxide

Synonyms: Sand; white quartz; quartz; silicon dioxide.

Formula: SiO₂
Boiling Pt: 2,210 °C (1.013 hPa)
Melting Pt: 1,710 °C
Density: 2,650 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 14808-60-7
EINECS: 238-878-4

Order Now

Specification Test Results

Particle Size 0.5-10 μm (approx. 80% between 1-5 μm)
Assay Min. 98 %
Grain size < 8 micron Min. 97 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR