Single-use syringes, 2-piece, HENKE-JECT®
Supplier: HENKE SASS WOLF
4010.200V0 4050.000V0 4020.000V0 4010.100V0 4010.A00V0 4020.X00V0 4050.X00V0 4100.000V0 4100.X00V0 4200.000V0 4200.X00V0 4050.090D0 4100.090D0 4200.090D0 4100X90D0 4050.X90D0 8300020452 8300020455 8300020456 8300020457 8300020458
HSWA4010.200V0EA 19.3 SGD
HSWA4010.200V0 HSWA4050.000V0 HSWA4020.000V0 HSWA4010.100V0 HSWA4010.A00V0 HSWA4020.X00V0 HSWA4050.X00V0 HSWA4100.000V0 HSWA4100.X00V0 HSWA4200.000V0 HSWA4200.X00V0 HSWA4050.090D0 HSWA4100.090D0 HSWA4200.090D0 HSWA4100X90D0 HSWA4050.X90D0 HSWA8300020452 HSWA8300020455 HSWA8300020456 HSWA8300020457 HSWA8300020458
Single-use syringes, 2-piece, HENKE-JECT®
Syringes Syringes, Medical
Disposable, two-piece, without needle, PP barrel and PE piston, latex- and silicone oil-free. Syringes are also available with especially designed oral tip, which prevents accidental Luer or hypodermic connection, in volumes of 5 or 10 ml. Incompatible with injection needles. Please enquire for details.
  • Defined position of the plunger at volume '0' to feel when plunger is completely inserted
  • No dead space for 1 ml insulin and tuberculin syringe
  • Extended graduation for wider range of applications
  • Positive safety stop to prevent accidental spills
Order Now
SPECIFICATIONS
Environmentally Preferable