Single-use syringes, 2-piece, HENKE-JECT®
Supplier: HENKE SASS WOLF
4010.200V0 4050.000V0 4020.000V0 4010.100V0 4010.A00V0 4020.X00V0 4050.X00V0 4100.000V0 4100.X00V0 4200.000V0 4200.X00V0 4050.090D0 4100.090D0 4200.090D0 4100X90D0 8300020455 8300020456 8300020458 8300063458 8300063457 8300065161 8300065162 8300063464 8300065163 8300063473 8300063460 8300063459 8300063466 8300065164 8300063461 8300065166 8300065230 8300065231 8300065229 8300065225 8300065234 8300065228
HSWA4010.200V0EA 19.3 SGD
HSWA4010.200V0 HSWA4050.000V0 HSWA4020.000V0 HSWA4010.100V0 HSWA4010.A00V0 HSWA4020.X00V0 HSWA4050.X00V0 HSWA4100.000V0 HSWA4100.X00V0 HSWA4200.000V0 HSWA4200.X00V0 HSWA4050.090D0 HSWA4100.090D0 HSWA4200.090D0 HSWA4100X90D0 HSWA8300020455 HSWA8300020456 HSWA8300020458 HSWA8300063458 HSWA8300063457 HSWA8300065161 HSWA8300065162 HSWA8300063464 HSWA8300065163 HSWA8300063473 HSWA8300063460 HSWA8300063459 HSWA8300063466 HSWA8300065164 HSWA8300063461 HSWA8300065166 HSWA8300065230 HSWA8300065231 HSWA8300065229 HSWA8300065225 HSWA8300065234 HSWA8300065228
Single-use syringes, 2-piece, HENKE-JECT®
Syringes Syringes, Medical
Disposable, two-piece, without needle, PP barrel and PE piston, latex- and silicone oil-free. Syringes are also available with especially designed oral tip, which prevents accidental Luer or hypodermic connection, in volumes of 5 or 10 ml. Incompatible with injection needles. Please enquire for details.
  • Defined position of the plunger at volume '0' to feel when plunger is completely inserted
  • No dead space for 1 ml insulin and tuberculin syringe
  • Extended graduation for wider range of applications
  • Positive safety stop to prevent accidental spills