Single-use syringes, 3-piece, HENKE-JECT®
Supplier: HENKE SASS WOLF
5010.200V0 5010.A00V0 5100.000V0 8300005761 8300005762 5050.000V0 5050.X00V0 5100.X00V0 5200.000V0 5200.X00V0 8300006682 8300006681 8300006680 8300005458 5100.090D0 5100.X90D0 8300014579 8300019869 8300005157 5300.000D0 5300.X00D0 8300063477 8300065154 8300063469 8300063470 8300063472 8300065151 8300065156 8300065153 8300065157 8300065152 8300063471 8300063465
HSWA5010.200V0EA 21 SGD
HSWA5010.200V0 HSWA5010.A00V0 HSWA5100.000V0 HSWA8300005761 HSWA8300005762 HSWA5050.000V0 HSWA5050.X00V0 HSWA5100.X00V0 HSWA5200.000V0 HSWA5200.X00V0 HSWA8300006682 HSWA8300006681 HSWA8300006680 HSWA8300005458 HSWA5100.090D0 HSWA5100.X90D0 HSWA8300014579 HSWA8300019869 HSWA8300005157 HSWA5300.000D0 HSWA5300.X00D0 HSWA8300063477 HSWA8300065154 HSWA8300063469 HSWA8300063470 HSWA8300063472 HSWA8300065151 HSWA8300065156 HSWA8300065153 HSWA8300065157 HSWA8300065152 HSWA8300063471 HSWA8300063465
Single-use syringes, 3-piece, HENKE-JECT®
Syringes Syringes, Medical
Disposable, three-piece syringes with PP barrel and piston, polyisoprene stopper, latex-free.
  • Lubricated synthetic rubber piston allows a smooth draw
  • Available in non sterile version (bulk) upon request
Without needles: Luer lock types are compatible for use with most common syringe pumps. Please ask for further information.
With fixed needles: Needle tube permanently attached in syringe, no dead space, safety stop, suitable for dosage of very small volumes.
Luer lock types are compatible for use with most common syringe pumps. Please ask forMore Product Information