Super elastic plunger microlitre syringes
Supplier: Hospidex
EX-MSE05 EX-MSE10 EX-MSNE05 EX-MSNE10 EX-MSE05R EX-MSNE05R EX-MSNE10R EX-MSE05WTS EX-MSE10WTS EX-MSNE05WTS EX-MSNE10WTS
HUAREX-MSE05EA 168 SGD
HUAREX-MSE05 HUAREX-MSE10 HUAREX-MSNE05 HUAREX-MSNE10 HUAREX-MSE05R HUAREX-MSNE05R HUAREX-MSNE10R HUAREX-MSE05WTS HUAREX-MSE10WTS HUAREX-MSNE05WTS HUAREX-MSNE10WTS
Super elastic plunger microlitre syringes
Syringes Syringes for Chromatography
Super elastic plunger is made of special Ti-Ni alloy which has superior flexibility and it does not have any problem with friction damage caused by plunger bending. It simply springs back, if the plunger is bent.
  • Superior chemical resistance
  • Plunger can be wound into a coil of 16 mm Ø
  • Accuracy of injection is about ±1%
  • Provided with 3-coloured index labels