Sterile Goods Baskets for ISO Units
Supplier: HUPFER METALLWERKE
7214006
HUPF7214006EA 428 SGD
HUPF7214006
Sterile Goods Baskets for ISO Units
Baskets Wire Baskets
Sterile supplies basket with insert frame for transporting, storing, and presenting sterile supplies in soft packaging without the use of containers.
  • Produced from electro-polished stainless steel
  • With U-shape profile rail compatible drawer frame
  • Stackable inside and on top of each other
Basket suitable for washing and sterilising devices, made from premium electro-polished round stainless steel material. Basket with upper insert frame suitable for sliding into L or U profile rails of cabinet systems, Norm 5 Medidul shelves or Hupfer Känguruh System®.More Product Information