Wall mounted hook rails
Supplier: HUPFER METALLWERKE
7500027 7500186 7500031 7500189 7500190 7500188 7500187 7500029
HUPF7500027EA 148 SGD
HUPF7500027 HUPF7500186 HUPF7500031 HUPF7500189 HUPF7500190 HUPF7500188 HUPF7500187 HUPF7500029
Wall mounted hook rails
Furniture Shelves
These numerous wallmounted hook rails with varied dimensions will meet all the requirements according to the type of material to be stored on the walls of storage areas.
  • High quality stainless steel square tube (25×25 mm) with welded hooks
  • Hook loading vertical 60 kg each hook, bolt for equipotential bonding
  • Tube ends covered with plastic caps
  • Supports are equipped with anti-static connecting pins and the screw heads are not visible
They offer the advantage of being able to store all the types of baskets at a place clearly defined in advance. They doMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable