Trolleys, basket, KR 60-60
Supplier: HUPFER METALLWERKE
7500196
HUPF7500196EA 2010 SGD
HUPF7500196
Trolleys, basket, KR 60-60
Trolleys
These trolleys made of stainless steel can take baskets, containers and bins made from polypropylene.
  • Stable, mobile round tube construction
  • Mobile on four swivel castors, two of them with brake
  • Galvanised-chromed housings with electro-conductive double ball-bearing
  • Two bumpers protect trolley against damage