Equipment table
Supplier: HUPFER METALLWERKE
7500204
HUPF7500204EA 1970 SGD
HUPF7500204
Equipment table
Furniture Tables and Workbenches
The instrument table is made from stainless steel. It is a solid, jointless tube structure made from square-type tube (20×20 mm), with two removable table tops formed as protection gallery (border top).
  • Electro-conductive
  • With double ball bearing the thread protection
  • Castors with 75 mm diameter
The instrument table is to provide immersion trays, sterile goods containers and baskets in operating theatres and neighbouring areas.
Certifications: Conforms with DIN 90952 packaging for sterilising goods.