Wire baskets
Supplier: HUPFER METALLWERKE
7500900 7500901 7500902 7500903 7500904 7500905 7500906 7500907 7500929
HUPF7500900EA 129 SGD
HUPF7500900 HUPF7500901 HUPF7500902 HUPF7500903 HUPF7500904 HUPF7500905 HUPF7500906 HUPF7500907 HUPF7500929
Wire baskets
Baskets Wire Baskets
Stainless steel.
  • Spot welded mesh 8×8×1 mm
  • Electrolytically polished
  • Reinforced edges
These baskets can be used for transport, storage, cleaning and sterilisation of laboratory utensils.
Order Now
SPECIFICATIONS
Environmentally Preferable