Wire basket
Supplier: HUPFER METALLWERKE
7500908
HUPF7500908EA 635 SGD
HUPF7500908
Wire basket
Baskets Wire Baskets
Stainless steel, electrolytically polished.
  • Very good chemical and thermal resistance
  • Foldaway carrying handle with plastic handgrip
  • Robust design (wire mesh size 8×8 mm) with two rods for base reinforcement
This rectangular baskets are suitable for a variety of uses.