Transport trolleys
Supplier: HUPFER METALLWERKE
7504001 7504009 7504107 7504002 7504006 7504005 7504128 7504114 7504129 7504110
HUPF7504001EA 1260 SGD
HUPF7504001 HUPF7504009 HUPF7504107 HUPF7504002 HUPF7504006 HUPF7504005 HUPF7504128 HUPF7504114 HUPF7504129 HUPF7504110
Transport trolleys
Trolleys
Transport trolley with high quality stainless steel construction, complete with 25 mm deep shelves. Shelves have bevelled edges for user comfort.
  • Bumper-type wall deflectors
  • Anti-static tyres
  • Chassis complete with four swivel castors, two front castors are lockable
Order Now
SPECIFICATIONS
Environmentally Preferable