Worktables
Supplier: HUPFER METALLWERKE
7504583
HUPF7504583EA 5800 SGD
HUPF7504583
Worktables
Furniture Tables and Workbenches
This open version work table features closed table top and bottom side covered with 0,8 mm sheet steel.
  • Particle board stuffing
  • Sandwich panelled table top
Working table made of stainless steel with grain size 400 ground and material thickness of 1 mm.