Cation exchange HPLC columns, ProPac™ SCX-10 and WCX-10
Supplier: Thermo Fisher Scientific
054993
HYPE054993EA 4040 SGD
HYPE054993
Cation exchange HPLC columns, ProPac™ SCX-10 and WCX-10
Chromatography Columns
The ProPac HPLC columns are weak and strong cation exchange columns with exceptionally high resolution and efficiency for separations of protein variants.
  • Characterisation and quality control assessment of monoclonal antibodies and other proteins
  • Useful for characterisation of related protein variants including deamidation and monoclonal antibodies lysine truncation variants
Based on stationary phases composed of 10 µm nonporous polymeric beads, these columns resolve isoforms that differ by a single charged residue. A thin, hydrophilic layer graftedMore Product Information