HPLC columns, Acclaim™ mixed-mode WCX-1
Supplier: Thermo Fisher Scientific
068353
HYPE068353EA 0 SGD
HYPE068353
HPLC columns, Acclaim™ mixed-mode WCX-1
Chromatography Columns
These are high-efficiency, silica-based columns, manufactured by bonding a specially designed proprietary ligand with both hydrophobic and weak cation-exchange properties. Selectivity of ionisable and neutral compounds can be controlled independently or simultaneously by tuning mobile phase ionic strength, pH or organic modifier.
  • Adjustable selectivity
  • Selectivity complementary to RP columns
  • Ideal selectivity for separating basic molecules
  • Multimode separation mechanism: RP, cation-exchange, ion-exclusion and HILIC
  • Capable of separating a mixture of
More Product Information