Aluminium oxide, Ecochrom™ Alumina B Activity: Super I

Supplier: MP Biomedicals

Synonyms: Aluminium(III) oxide, Alumina C, Aluminum oxide

0204571
ICNA0204571EA 413 SGD
ICNA0204571
Aluminium oxide, Ecochrom™ Alumina B Activity: Super I
Aluminium oxide

Specific Surface Area: 200 m2/g
Particle Size: 63-200 µm
Weight: 0.8 g/ml pH: 10


pH: 10

Formula: Al₂O₃
Density: 3,94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR