Agarose for electrophoresis

Supplier: MP Biomedicals

0210026790 0210026705 0210026710 0210026725 0210026750 0210026780 0210026783
ICNA0210026790EA 2200 SGD
ICNA0210026790 ICNA0210026705 ICNA0210026710 ICNA0210026725 ICNA0210026750 ICNA0210026780 ICNA0210026783
Agarose for electrophoresis
Agarose
MDL Number: MFCD00081294
CAS Number: 9012-36-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=526c20a29610acf6900f898ff245c5602558c785&vid=ae34e4c76a4d7838d4dc9501397f6741b1b97d5e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Synonyms (1→4)-3,6-anhydro-α-L-galactopyranosyl-(1→3)-β-D-galactopyranan; 3,6-Anhydro-a-L-galacto-b-D-galactan; FastLane agarose; Indubiose A4; NuSieve GTG; Odigose; Seakem; Sepharose
Form White Powder
Storage Store at Room Temperature (15 - 30 °C)
Electroendosmosis 0,11 - 0,17
Gel Strength ≥2000 g/cm2 (1,5% gel)
Gel Temperature 35 - 37 °C
Moisture Content ≤5%
pH 5,5 - 7,5 (1% aq soln)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR