Alizarin red S (sodium salt), anhydrous, powder_orange C.I. 58005

Supplier: MP Biomedicals
Warning

Synonyms: Sodium 3,4-dihydroxy-9,10-dioxo-9,10-dihydroanthracene-2-sulphonate, C.I. 58005, Alizarin sulphonic acid disodium salt, Alizarin Red S sodium

0210037590 0210037525 0210037580
ICNA0210037590EA 2000 SGD
ICNA0210037590 ICNA0210037525 ICNA0210037580
Alizarin red S (sodium salt), anhydrous, powder_orange C.I. 58005
Alizarin Red S (sodium salt)

Alizarin red S is used in a biochemical assay to determine, quantitatively by colorimetry, the presence of calcific deposition by cells of an osteogenic lineage.


Alizarin red S is used in a biochemical assay to determine, quantitatively by colorimetry, the presence of calcific deposition by cells of an osteogenic lineage.


Soluble in water (1 mg/ml).

Formula: C₁₄H₇NaO₇S
MW: 342.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00013049
CAS Number: 130-22-3
EINECS: 204-981-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR