L-α-Glycerophosphoryl Choline, Cadmium Chloride Complex (from egg yolk) ∼98%, white powder

Supplier: MP Biomedicals
Danger

Synonyms: sn-Glycero-3-phosphocholine 1:1 cadmium chloride adduct, α-GPC, Choline, Cadmium Chloride Complex

02100487.5 0210048701
ICNA02100487.5EA 0 SGD
ICNA02100487.5 ICNA0210048701
L-α-Glycerophosphoryl Choline, Cadmium Chloride Complex (from egg yolk) ∼98%, white powder
L-α-Glycerophosphoryl Choline, Cadmium Chloride Complex
Formula: C₈H₂₀CdCl₂NO₆P
Storage Temperature: Freezer
CAS Number: 64681-08-9
EINECS: 265-010-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR