DL-Arabinose, white powder

Supplier: MP Biomedicals

0210070010
ICNA0210070010EA 234 SGD
ICNA0210070010
DL-Arabinose, white powder
DL-Arabinose

DL-Arabinose is a mixture of the diastereomers D-arabinose and L-arabinose


Caution: Research Use Only (RUO).

Formula: C₅H₁₀O₅
Storage Temperature: Ambient
MDL Number: MFCD00135867
CAS Number: 147-81-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR