β-Alanine ≥98%, crystalline powder

Supplier: MP Biomedicals

Synonyms: H-β-Ala-OH, 3-Aminopropionic acid, H-Bala-OH, 3-Aminopropanoic acid

02100929.5 0210092901
ICNA02100929.5EA 0 SGD
ICNA02100929.5 ICNA0210092901
β-Alanine ≥98%, crystalline powder
β-Alanine

β-Alanine is an N-blocked form of alanine. A nonselective endogenous glycine receptor agonist; a taurine analogue; an electrolyte.


β-Alanine is used in the synthesis of pantothenic acid and derivatives; as buffer in electroplating.


β-Alanine, a β -amino acid, is a component of pantothenic acid and the rate-limiting amino acid in the biosynthesis of the histidinyl antioxidant dipeptides carnosine and anserine. Endogenous β-amino acid that is a nonselective agonist at glycine receptors and a ligand for the G protein-coupled orphan receptor, TGR7 (MrgD). β-Alanine flux plays a cytoprotective role by supporting the osmotic stability of marine organisms, preimplantation mouse embryos and mammalian cells exposed to hypoxic stress.


Soluble in water (100 mg/ml - clear, colorless solution).


Caution: Research Use Only (RUO).

Formula: C₃H₇NO₂
Melting Pt: 200 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00008200
CAS Number: 107-95-9

Order Now

Specification Test Results

Identity Test Passes
Purity ≥98%
FTIR Conforms to
Standard
Loss on Drying ≤0,6%
Residue on Ignition ≤0,2%
Ammonia 0,1%
Chloride 0,05%
Sulphate 0,05%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR