Crotalus adamanteus Phosphodiesterase I, MP Biomedicals

Supplier: MP Biomedicals

0210097880
ICNA0210097880EA 578 SGD
ICNA0210097880
Crotalus adamanteus Phosphodiesterase I, MP Biomedicals
Enzymes

Lyophilised in vials containing approximately 5 mg dry wt. and at least 100 units/vial (20 to 40 units/mg dry wt.).


Unit definition: One unit hydrolyses 1 µmole of p-nitrophenyl thymidine-5-phosphate per minute at pH 8,9 and 25 °C


pH: optimum pH=9,8 to 10,4.5 (Lit.)

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR