Deoxycholic acid Free_acid 99, powder_white

Supplier: MP Biomedicals

Synonyms: 12α-Hydroxylithocholic acid, (R)-4-((3R,5R,8R,9S,10S,12S,13R,14S,17R)-3,12-Dihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanoic acid, 3α,12α-Dihydroxy-5β-cholanic acid, 3α,12α-Dihydroxy-5β-cholan-24-oic acid, DCA, 12alpha-Hydroxylithocholic acid, 3α,12α-Dihydroxycholan-24-oic acid, 3α,12α-Dihydroxycholanic acid

0210149610 0210149680
ICNA0210149610EA 107 SGD
ICNA0210149610 ICNA0210149680
Deoxycholic acid Free_acid 99, powder_white
Deoxycholic acid

Deoxycholic acid is a bile acid formed by metabolism of cholate.


Deoxycholic acid has been used in a modified procedure to recover 40 to 80% of a protein from a 1 μg/ml solution. It forms complexes with fatty acid.


The free acid is soluble at 0,24 g/L in water at 15 °C.


Caution: Research Use Only (RUO).

Formula: C₂₄H₄₀O₄
MW: 392.58
Boiling Pt: 504
Melting Pt: 190
Storage Temperature: Ambient
MDL Number: MFCD00003673
CAS Number: 83-44-3
Merck Index: 13,02916

Order Now

Specification Test Results

Identity Test Passes
Purity ≥99%
FTIR Conforms to Standard
Melting Point 168 - 178 °C
Extinction Coefficient EmM = 9,4 (Lit.)
Optical Rotation +54,1° (c=2 in MeOH) (Lit.)
pka 6,58 (Lit.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR