Hydrocortisone-21-acetate

Supplier: MP Biomedicals
Warning

Synonyms: Hydrocortisone acetate

0210199505 0210199510
ICNA0210199505EA 787 SGD
ICNA0210199505 ICNA0210199510
Hydrocortisone-21-acetate
Hydrocortisone 21-acetate

HYDROCORTISONE-21-ACETATE (Δ4-Pregnen-11β,17α,21-triol- 3,20-dione; Cortisol acetate) Crystalline

Formula: C₂₃H₃₂O₆
MW: 404.51 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00037714
CAS Number: 50-03-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR