D-(-)-Fructose, powder

Supplier: MP Biomedicals

Synonyms: D(-)-Levulose, (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one, β-D-(-)-Fructose, Fructose

02102186.5
ICNA02102186.5EA 599 SGD
ICNA02102186.5
D-(-)-Fructose, powder
D(-)-Fructose

Fructose or fruit sugar, is a simple monosaccharide found in many plants. It is one of the three dietary monosaccharides, along with glucose and galactose, that are absorbed directly into the bloodstream during digestion. Fructose is a 6-carbon polyhydroxyketone. It is an isomer of glucose which can be anaerobically fermented by yeast or bacteria.


  • Presentation: Powder


Fructose and other monosaccharides are used for the diagnosis of cancer as cancer cells have shown a higher level of fructose utilisation as compared to glucose.


Soluble in water and alcohols.

Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Boiling Pt: 552
Melting Pt: 110…120 °C
Density: 1.6 g/mL (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00148910
CAS Number: 57-48-7
Merck Index: 13,04295
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d8c96c34b23753d10b06108938876644c747995c&vid=6d952afc3154d3befcf243a015f513a9130c0835&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Purity ≥99%
FTIR Conforms to Standard
Optical Rotation (c=2, water) -88° to -98°
Arsenic <1 ppm
Heavy Metals <5 ppm
Moisture (KF) 0.02%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR